Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
US FDA LIB 4423: GC Analysis of Melamine and Related Compounds in Dog Food on SLB®-5ms
US FDA LIB 4423: GC Analysis of Melamine and Related Compounds in Dog Food on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 115 °C (3 min), 10 °C/min to 325 °C (6 min)
carrier gas helium, 1 mL/min, constant
sample TMS derivatives of melamine and related compounds (extract of dog food, with I.S. added at 1000 ng/mL, in pyridine:sylon BFT)
injection 1 µLsplitless
liner 4 mm I.D. single taper
inj. temp. 250 °C
MSD interface 325 °C
scan range SIM
Description
Categories Analytical Chromatography, GC Applications, Amines
Featured Industry Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 2,6-Diamino-4-chloropyrimidine
  • Ammelide
  • Ammeline
  • Cyanuric acid
  • Melamine