Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
AOAC Method 2011.11: UHPLC/MS/MS Analysis of Vitamin D in Milk on Titan™ C18
AOAC Method 2011.11: UHPLC/MS/MS Analysis of Vitamin D in Milk on Titan™ C18
Conditions
column Titan C18, 10 cm x 2.1 mm I.D., 1.9 µm particles (577124-U)
column temp. 25 °C
mobile phase [A] 0.1% formic in methanol:water (20:80); [B] 0.1% formic acid in methanol
gradient 60 to 90% B in 0.4 min; to 100% B in 0.3 min; held at 100% B for 7.8 min; to 60% B in 0.1 min; held at 60% B for 1.5 min
flow rate 0.25 mL/min for 5.55 min; to 0.50 mL/min in 0.1 min; held at 0.5 mL/min for 3.84 min; to 0.25 mL/min in 0.1 min; held at 0.25 mL/min for 0.7 min
pressure 5859 psi (404 bar)
sample 1 IU/mL in methanol
injection 5 µL
detector MS, APCI(+), MRM 397/125 (ergocalciferol, D2), MRM 385/259 (cholecalciferol, D3)
Description
Analysis Note AOAC method 2011.11 describes the procedure the analysis of vitamin D in infant formula and adult/pediatric nutritional formula using UHPLC/MS/MS. Two forms of vitamin D are recognized, vitamin D2 or ergocalciferol, and vitamin D3 or cholecalciferol. The AOAC method was followed in the work presented here to measure vitamin D in milk using a Titan C18 UHPLC column. Vitamin D3 and very low levels of D2 were detected,
Categories Analytical Chromatography, Food & Beverage Analysis, Beverage Testing, Milk
Featured Industry Food and Beverages
Legal Information Titan is a trademark of Sigma-Aldrich Co. LLC
suitability application for UHPLC