Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Effect of Mobile Phase Ionic Component on the HPLC Analysis of the Photosensitizer Aminolevulinic Acid on SeQuant ZIC-HILIC
Effect of Mobile Phase Ionic Component on the HPLC Analysis of the Photosensitizer Aminolevulinic Acid on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 50 x 4.6 mm, 5 µm, 200 Ã… (1.50451.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water or ammonium acetate or ammonium formate; (70:30, A:B)
flow rate 0.5 mL/min
pressure 662 psi (4.6 MPa)
injection 5 µL
detector 254 nm
Description
Categories Analytical Chromatography, Drugs, miscellaneous