Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
ELSD/MS Analysis of Choline and Inositol on SeQuant ZIC-HILIC
ELSD/MS Analysis of Choline and Inositol on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 20 x 2.1 mm, 3.5 µm, 100 Ã… (1.50439.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate; (80:20, A,B)
flow rate 0.5 mL/min
pressure 792 psi (5.5 MPa)
injection 2 µL
detector Sedex 85LT ELSD @ 60 ºC, 4.0 bar compressed air and Gain 11
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products