Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
High Efficiency Resolution on Ascentis® Express 2.7 μm Versus 3μm Particles
High Efficiency Resolution on Ascentis® Express 2.7 μm Versus 3μm Particles
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 µm particles and C18, 15 cm x 4.6 mm I.D., 3 µm particles (53829-U)
column temp. ambient
mobile phase [A] water; [B] acetontrile; (35:65 or 27.5:72.5, A:B)
flow rate 1.5 mL/min
injection 2 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Semivolatiles