Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of (+/-)-N,N-Dibenzyltartaric Diamide Enantiomers on Astec® (R,R) P-CAP™-DP
HPLC Analysis of (+/-)-N,N-Dibenzyltartaric Diamide Enantiomers on Astec® (R,R) P-CAP™-DP
Conditions
column Astec (R,R) P-CAP-DP, 25 cm x 4.6 mm I.D., 5 µm particles (35024AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] methanol; (95:5, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 1 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Bases, Chemicals and Industrial Polymers, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information P-CAP is a trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC