Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of 2(2-Chlorophenoxy) Propionic Acid Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of 2(2-Chlorophenoxy) Propionic Acid Enantiomers on Astec® CHIROBIOTIC® TAG
Conditions
column CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 µm particles (14024AST)
column temp. 25 °C
mobile phase 0.1 wt% ammonium acetate in methanol
flow rate 1 mL/min
sample 2 mg/mL in methanol
injection 2 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Environmental, Pesticides & Herbicides, All Environmental Applications, Herbicides
Featured Industry Agriculture
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC