HPLC Analysis of 3and 4Amino3Hydroxyphenylalanine (3AHP and 4AHP) on SeQuant® ZICHILIC
HPLC Analysis of 3and 4Amino3Hydroxyphenylalanine (3AHP and 4AHP) on SeQuant® ZICHILIC
Conditions
column SeQuant ZICHILIC, PEEK, 150 x 2.1 mm, 3.5 μm, 100 Å (1.50442.0001)
mobile phase [A] acetonitrile; [B] ammonium acetate 100mM; (75:25, A:B)
flow rate 0.15 mL/min
pressure 936 psi (6.5 MPa)
detector 254 nm
injection 1 μL
Description
Categories Amino Acids, Analytical/Chromatography, Applications, HPLC Applications
Legal Information SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany