HPLC Analysis of a Procainamide-Labeled Dextran Ladder on BIOshell™ Glycan using HILIC-FLR
HPLC Analysis of a Procainamide-Labeled Dextran Ladder on BIOshell™ Glycan using HILIC-FLR
Conditions
column HPLC BIOshell Glycan Column, 15 cm x 2.1 mm I.D., 2.7 µm particles (50994-U)
mobile phase A: 50 mM ammonium formate (Product No. 17843) pH 4.4, [ammonium] = 50 mM, pH adjust with formic acid B: Acetonitrile (Product No. 34851)
gradient 75% to 59% B in 75 min
flow rate 0.3 mL/min
sample Dextran Ladder (Product 31417) after Procainamide Labeling Protocol
injection 10 µL
detector FLR (308 nm excitation, 359 emission)
Description
Analysis Note The dextran ladder is used as an external standard for the analysis of glycans by normal phase HPLC after fluorescent labeling. When analyzed on the BIOshell Glycan HPLC Column, this standard gives a characteristic ladder profile from monomeric glucose to approximately a 20-mers of glucose oligosaccharide. This ladder provides calibration reference points that can aide in identifying more complex glycans based upon retention characteristics.
Categories Analytical Chromatography, Proteins, Peptides