Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Acidic and Neutral Drug Panel on Ascentis® Express C18
HPLC Analysis of Acidic and Neutral Drug Panel on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 4.6 mm I.D., 2.7 µm particles (53827-U)
column temp. ambient
mobile phase [A] water with 0.1% phosphoric acid, [B] acetonitrile with 0.1% phosphoric acid
flow rate 1.76 mL/min
injection 100 µL
detector UV, 215 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Hormones, Anticonvulsant/antiepiletic/antiseizure, Anti-inflammatories, Antihypertensives