HPLC Analysis of Acidic Drugs on Chromolith(R) Performance 100- 2 mm
HPLC Analysis of Acidic Drugs on Chromolith® Performance 100- 2 mm
Conditions
column Chromolith® Performance 100 x 2 mm (1.52006.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] ammoniumacetate 20mM pH=6,8; (28:70, A:B)
flow rate 380 μL/min (u=2mm/s)
pressure 461 psi (32 bar)
sample Indoprofen, Naprofen, Ethacrynic acid, Ibuprofen, Indomethacine
injection 1.0 μL
detector 230 nm (cell: 1.4 μL)
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany