Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Acidic Drugs on Chromolith(R) Performance 100- 3 mm
HPLC Analysis of Acidic Drugs on Chromolith® Performance 100- 3 mm
Conditions
column Chromolith® Performance 100 x 3 mm (1.52001.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] ammoniumacetate 20mM pH=6,8; (28:70, A:B)
flow rate 850 μL/min (u=2mm/s)
pressure 5196 psi (36 bar)
sample Indoprofen, Naprofen, Ethacrynic acid, Ibuprofen, Indomethacine
injection 1.0 μL
detector 230 nm cell: 1.4 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany