HPLC Analysis of Acidic Drugs on Chromolith(R) Performance 100- 4 mm (analyte set 1
HPLC Analysis of Acidic Drugs on Chromolith® Performance 100- 4 mm (analyte set 1
Conditions
column Chromolith® Performance 100 x 4 mm (1.52028.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] ammoniumacetate 20mM pH=6,8; (28:70, A:B)
flow rate 2 mL/min (u=2mm/s)
pressure 14001 psi (97 bar)
sample Indoprofen, Naprofen, Ethacrynic acid, Ibuprofen, Indomethacine
injection 1 μL
detector 230 nm (cell: 1.4 μL)
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany