HPLC Analysis of Albuterol Tablets - Assay on Chromolith(R) HighResolution RP-18 endcapped 150 x 4.6mm
HPLC Analysis of Albuterol Tablets - Assay on Chromolith® HighResolution RP-18 endcapped 150 x 4.6mm
Conditions
sample preparation 120 μg/mL of Albuterol Sulfate RS prepared as follows. Transfer USP Albuterol Sulfate RS to a suitable volumetric flask, and add a volumne of Solution A corresponding to 60% of the flask volume. Sonicate for 5 minutes, and dilute with Methanol to volume.
column Chromolith® HighResolution RP-18 endcapped, 150 x 4.6 mm (1.52023.0001)
column temp. ambient
mobile phase [A] 10mL/L of glacial acetic acid in water; [B] 1.13g of sodium 1-Hexanesulfate in 1200mL water, add 12mL of glacial acetic acid; (40:60,:A:B)
flow rate 1.5 mL/min
pressure 2001 psi (138 bar)
sample Albuterol RS 30 μg/mL in diluent (30 μg/mL of Albuterol Sulfate RS in Diluent from Standard stock solution)
injection 25 μL
detector 276 nm cell: 11 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany