Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Alkaloids from Herbal Supplements on Ascentis® Express F5, HILIC Mode
HPLC Analysis of Alkaloids from Herbal Supplements on Ascentis® Express F5, HILIC Mode
Conditions
column Ascentis® Express F5, 3 cm x 3.0 mm I.D., 2.7 µm particles (53574-U)
column temp. 35 °C
mobile phase 2 mM ammonium acetate in 10:90 water:acetonitrile, pH to 6.0 with acetic acid
flow rate 0.5 mL/min
pressure 1100 psi (76 bar)
sample methanolic extract of Yohimbe Herbal Supplement
injection 2 µL
detector UV, 275 nm
Description
Categories Analytical Chromatography, ED Drugs
Featured Industry Vitamins, Nutraceuticals, and Natural Products