Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Aminopterin and Related Compounds on Ascentis® Express C18
HPLC Analysis of Aminopterin and Related Compounds on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 µm particles (53829-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate, pH 3.0; [B] acetonitrile; (90:10, A:B)
flow rate 1.0 mL/min
injection 5 µL
detector UV, 280 nm
Description
Analysis Note This application demonstrates the suitability of the Ascentis C18 for the analysis of aminopterin, folic acid and methotrexate. Structures along with the optimized chromatogram obtained on the Ascentis Express C18 are presented
Categories Analytical Chromatography, Antineoplastics