HPLC Analysis of Amlodipine Besylate on Purospher(R) STAR RP-18 endcapped
HPLC Analysis of Amlodipine Besylate on Purospher® STAR RP-18 endcapped
sample preparation Dissolve appropriate amount of standard in mobile phase to obtain 50 μg/mL (50 ppm).
column Purospher® STAR RP-18 endcapped 250 x 4.6 mm (1.51456.0001)
column temp. 30 °C
mobile phase [A] methanol; [B] acetonitrile; [C] 0.7% triethylamine aqueous solution ; (35:15:50, A:B:C)
flow rate 1 mL/min
sample 50 ppm Amlodipine Standard Solution
injection 10 μL
detector 237 nm 1 μL/10 mm
Featured Industry Pharmaceutical (small molecule)
Legal Information Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany