Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Analgesics on Discovery® RP-Amide C16
HPLC Analysis of Analgesics on Discovery® RP-Amide C16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. ambient
mobile phase [A] 25 mM potassium phosphate (pH 2.3); [B] acetonitrile
flow rate 2 mL/min
injection 10µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Analgesics
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC