HPLC Analysis of Analgesics on Discovery® HS F5
HPLC Analysis of Analgesics on Discovery® HS F5
Conditions
column Discovery HS F5, 15 cm x 4.6 mm I.D., 5 µm particles (567516-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate, 0.1% formic acid; [B] methanol
flow rate 1.5 mL/min
injection 10 µL
detector UV, 254nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Analgesics
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC