Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Anti-Inflammatory Drugs on Discovery® HS F5
HPLC Analysis of Anti-Inflammatory Drugs on Discovery® HS F5
Conditions
column Discovery HS F5, 15 cm x 4.6 mm I.D., 5 µm particles (567516-U)
mobile phase [A] 10 mM potassium phosphate (pH 6.8); [B] acetonitrile; (85:15, A:B)
flow rate 1 mL/min
injection 5 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anti-inflammatories
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC