Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Antianxiety Drugs on Ascentis® Phenyl
HPLC Analysis of Antianxiety Drugs on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] water; (40:60, A:B)
flow rate 1.0 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anticonvulsant/antiepiletic/antiseizure, Anxiolytics, Sedatives