Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Antibacterials on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Antibacterials on Chromolith® High Resolution RP-18e
Conditions
column Chromolith® High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. ambient
mobile phase [A] methanol; [B] phosphat buffer 50mM pH=3,0; (55:45, A:B)
flow rate 2 mL/min
sample Penicilin 10,61mg/100ml, Ampicilin 20,63mg/100ml
injection 1 μL
detector 210 nm cell: 11 μL
Description
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany