Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Aripiprazole and Dehydro Aripiprazole Using an Ascentis® Express C18 Column
HPLC Analysis of Aripiprazole and Dehydro Aripiprazole Using an Ascentis® Express C18 Column
Conditions
column Ascentis Express C18, 10 cm  x 3.0 I.D mm., 2.0 µm particles (50819-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate; pH 3.0 with formic acid [B] 0.1% formic acid in methanol
gradient 0 to 40% B in 0.5 min; to 95% B in 3.5 minutes; held at 95% B for 2 min
flow rate 0.5 mL/min
pressure 6500 psi (445 bar)
sample 50 µg/mL in 10% methanol
injection 2 µL
detector UV, 245 nm
Description
Analysis Note Aripiprazole is marketed as the atypical antipsychotic Abilify® for the treatment of schizophrenia, bipolar disorder and clinical depression. Dehydroapiprazole is the primary metabolite of Aripiprazole.
Categories Analytical Chromatography, Antidepressants
Featured Industry Pharmaceutical (small molecule)
Legal Information Ascentis is a registered trademark of Sigma-Aldrich Co. LLC, Abilify is a registered trademark of Otsuka Pharmaceutical Co.