Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Artificial Food Colorants on Chromolith® HighResolution RP-18 endcapped, UV 510 nm
HPLC Analysis of Artificial Food Colorants on Chromolith® HighResolution RP-18 endcapped, UV 510 nm
Conditions
column Chromolith HighResolution RP-18, 10 cm x 4.6 mm I.D., 2 µm particles (1.52022.0001)
column temp. 30 °C
mobile phase [A] 20mM ammonium acetate pH 6.8 in water; [B] methanol
gradient 5 to 50% B in 6 min; to 100% B in 4 min; held at 100% B for 1 min; to 5% B in 0.01 min
flow rate 1 mL/min
pressure 1526 psi (106 bar)
sample Amaranth 0.05 mg/mL, Carmine Red 0.05 mg/mL, Sunset Yellow 0.05 mg/mL, Fancy Red 0.05 mg/mL, Erythrosine 0.05 mg/mL in methanol:water (50:50)
injection 20 µL
detector UV, 510 nm
Description
Categories Analytical Chromatography, Dyes
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany