Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Artificial Food Colorants in Orange Soft Drink on Chromolith® HighResolution RP-18 endcapped
HPLC Analysis of Artificial Food Colorants in Orange Soft Drink on Chromolith® HighResolution RP-18 endcapped
Conditions
column Chromolith HighResolution RP-18, 10 cm x 4.6 mm I.D., 2 µm particles (1.52022.0001)
mobile phase [A] 20mM ammonium acetate pH 6.8 in water; [B] methanol
gradient 5 to 50% B in 6 min; to 100% B in 4 min; held at 100% B for 1 min; to 5% B in 0.01 min
flow rate 1 mL/min
pressure 1526 psi (106 bar)
injection 20 µL
detector UV, 428 nm
Description
Categories Analytical Chromatography, Dyes
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany
Other Notes 5 mL of the beverage was transferred to a 10 mL volumetric flask, sonicated for 5 minutes, then pure water was added to the volume. The solution was filtered by 0.45 μm PTFE syringe filter into a 2mL autosampler vial prior to injection.