Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Ascorbic acid on Chromolith(R) NH2 100 - 4.6mm
HPLC Analysis of Ascorbic acid on Chromolith® NH2 100 - 4.6mm
Conditions
column Chromolith® NH2, 100 x 4.6 mm (1.52028.0001)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] ammonium acetate 100mM pH=6.8; (90:10, A:B)
flow rate 4 mL/min
pressure 1566 psi (108 bar)
sample (Ascorbic acid 1mg/ml, Dehydroascorbic acid 0.5mg/ml)
injection 5 μL
detector 240 nm cell: 11 μL
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany