HPLC Analysis of B-Vitamins on Chromolith(R) NH2 100 - 4.6mm
HPLC Analysis of B-Vitamins on Chromolith® NH2 100 - 4.6mm
Conditions
column Chromolith® NH2, 100 x 4.6 mm (1.52028.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (20:80, A:B)
gradient 0-3min 1mL/min, 3,1-10min 2mL/min
pressure 290 psi 7-20 Bar
sample Riboflavin (8.8mg/10ml), Pyridoxin ( 5.5mg/10ml), Cyanocobalamin (5.2mg/10ml)
injection 2 μL
detector 280 nm cell: 11 μL
Description
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany