HPLC Analysis of B-Vitamins on Chromolith NH2 50 - 4.6mm
HPLC Analysis of B-Vitamins on Chromolith NH2 50 - 4.6mm
Conditions
column Chromolith® NH2 50 x 4.6 mm (1.52027.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (74:26, A:B)
flow rate 2 mL/min
pressure 218 psi (15 bar)
sample Riboflavin (8.8mg/10ml), Pyridoxin ( 5.5mg/10ml), Cyanocobalamin (5.2mg/10ml)
injection 0.3 μL
detector 280 nm
Description
Featured Industry Food and Beverages