Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of B-Vitamins on Chromolith NH2 25 - 4.6mm
HPLC Analysis of B-Vitamins on Chromolith NH2 25 - 4.6mm
Conditions
column Chromolith® NH2 25 x 4.6 mm (1.52026.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (74:26, A:B)
flow rate 2 mL/min
pressure 57 psi (3.9 bar)
sample Riboflavin (8.8mg/10ml), Pyridoxin ( 5.5mg/10ml), Cyanocobalamin (5.2mg/10ml)
injection 0.3 μL
detector 280 nm
Description
Featured Industry Food and Beverages