HPLC Analysis of Benalaxyl Enantiomers on Astec(R) Cellulose DMP in Normal Phase Mode
HPLC Analysis of Benalaxyl Enantiomers on Astec(R) Cellulose DMP in Normal Phase Mode
Description
Analysis Note Benalaxyl is a fungicide used to protect a variety of crops, including potatoes and grapes. It is a chiral compound, comprising two enantiomers. Enantioselective influences of chiral pesticides, like Benalaxyl, and their metabolites on ecosystems is an area of great importance. The possibility that only one enantiomer has the wanted properties and/or the other form an unwanted effect may exist. An analytical separation of the enantiomers is required to perform studies in these areas. The Astec Cellulose DMP column resolved the enantiomers of Benalaxyl under normal phase conditions.
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC