Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Beta Blockers on Ascentis® Express C18
HPLC Analysis of Beta Blockers on Ascentis® Express C18
Conditions
column Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.7 µm particles (53822-U)
column temp. 35 °C
mobile phase [A] 0.1% acetic acid in water; [B] 0.1% acetic acid in acetonitrile
flow rate 0.2 mL/min
injection 1 µL
detector ABI 3200 QT; ESI(+), MS/MS
instrument Agilent 1100
Description
Categories Analytical Chromatography, Antihypertensives