Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Beta Blockers on Ascentis® Express C18 using a 60 Second Gradient
HPLC Analysis of Beta Blockers on Ascentis® Express C18 using a 60 Second Gradient
Conditions
column Ascentis Express C18, 2 cm x 2.1 mm I.D., 5 µm particles (50507-U)
column temp. 40 °C
mobile phase [A] water/0.1% TFA; [B] acetonitrile/0.1% TFA
gradient 5-50% B in 60 seconds
flow rate 2 mL/min
sample 0.17-0.33 mg/mL in 71% water/29% acetonitrile
injection 0.5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Beta-blockers