HPLC Analysis of Bimatoprost on Chromolith(R) Performance 100-2mm
HPLC Analysis of Bimatoprost on Chromolith® Performance 100-2mm
Conditions
column Chromolith® Performance, 100 x 2 mm (1.52006.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water + 0,05% TFA v/v; [C] methanol; (30:60:10, A:B:C)
flow rate 380 μL/min
pressure 691 psi (48 bar)
sample Bimatoprost (1mg/mL), Bimatoprost free acid (1mg/mL)
injection 1 μL
detector 210 nm cell: 1.4 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany