Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Biotin Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Biotin Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 23 °C
mobile phase [A] 0.1% sodium phosphate, pH 4.1; [B] methanol; (60:40, A:B)
flow rate 1 mL/min
sample 4 mg/mL in methanol
injection 2 µL
detector UV, 210 nm
Description
Categories Analytical Chromatography, Chiral Applications, Life Science & Biopharma, Vitamins and Natural products, Food & Beverage, All Food & Beverage Applications, All Biochemical Applications, Vitamins & Coenzymes, Vitamins
Featured Industry Vitamins, Nutraceuticals, and Natural Products
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC