Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Biphenyles on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Biphenyles on Chromolith® High Resolution RP-18e
Conditions
column Chromolith® High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] water; (40:60, A:B)
flow rate 4.0 mL/min
sample Biphenyl-4,4′-ol 1,5mg/10ml, Biphenyl-2,2′-ol 7,0mg/10ml, Biphenyl-4-ol 1mg/10ml, Biphenyl-2-ol 1mg/10ml
injection 5 μL
detector 254 nm cell: 11 μL
Description
Featured Industry General Analytical
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany