Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Caffeine and Taurine on Ascentis® Express HILIC Column using Evaporative Light Scattering Detection (ELSD)
HPLC Analysis of Caffeine and Taurine on Ascentis® Express HILIC Column using Evaporative Light Scattering Detection (ELSD)
Conditions
column Ascentis Express HILlC, 5 cm x 3.0 mm I.D., 2.7 µm particles (53967-U)
column temp. 25 °C
mobile phase [A] 100 mM ammonium formate, pH 3.0 with formic acid; [B] acetonitrile; (10:90, A:B)
flow rate 0.6 mL/min
sample 1000 µg/mL in 10:90, water: acetonitrile
injection 2 µL
detector ELSD, 55 °C, 3.5 bar nitrogen
Description
Analysis Note This application demonstrates the suitability of Ascentis Express HILIC for the efficient separation of caffeine and taurine. Elution is by aqueous normal phase (ANP) as an increase in organic will increase retention.
Categories Analytical Chromatography, StiµLants, Organic Acids, Drugs of Abuse
Featured Industry Vitamins, Nutraceuticals, and Natural Products