Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Catechols and Resorcinols on Ascentis® RP-Amide
HPLC Analysis of Catechols and Resorcinols on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 30 °C
mobile phase [A] 20 mM phosphoric acid (pH 2.0 unadjusted): [B] acetonitrile; (75:25, A:B)
flow rate 1.5 mL/min
sample as indicated in 20 mM phosphoric acid (pH 2.0)
injection 25 µL
detector UV, 270 nm
Description
Categories Analytical Chromatography, Clinical, antiseptic/disinfectants, phenols, Phenols, catecholamines
Featured Industry Chemicals and Industrial Polymers
Clinical