HPLC Analysis of Cetirizine on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Cetirizine on Chromolith® High Resolution RP-18e
column Chromolith® High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. 30 °C
mobile phase [A] 4.6 gammonium dihydrogen phosphate in 1L MilliQ water. Adjust pH to 6.0 with triethyl amine. Add 500 mg of 1-octane sulfonate. Filter through 0.2 μM filter paper Mix Buffer & Tetrahydrofuran 90:10 (v/v); [B] Mix methano & iso-propanol 75:25 (v/v); Mix mobile phase A & B 60:40 (v/v)
flow rate 0.5 mL/min
pressure 624 psi (43 bar)
sample Weigh 25 mg substance in 50 ml volumetric flask. Dissove in diluent & make up the volume with diluent. (Diluent:Mix buffer & acetonitrile 75:25 (v/v))
injection 10 μL
detector 230 nm cell: 10 μL
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany