HPLC Analysis of Cetirizine Hydrochloride on Purospher(R) STAR RP-18 endcapped
HPLC Analysis of Cetirizine Hydrochloride on Purospher® STAR RP-18 endcapped
Conditions
column Purospher® STAR RP-18 endcapped 150 x 4.6 mm (1.51455.0001)
column temp. 30 °C
mobile phase [A] 100mM NaH2PO4 aqueous solution, adjusted to pH 3.7 with H3PO4; [B] acetonitrile; (60:40, A:B)
flow rate 1 mL/min
pressure 1465 psi 101 Bar (101 Bar)
sample Weigh appropriate amount of Cetirizine, add diluent to obtain 100 μg/mL (100 ppm) concentration.
injection 20 μL
detector 232 nm cell: 1 μL/10 mm
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany