HPLC Analysis of Chlorocresol on Chromolith(R) HighResolution RP-18 endcapped
HPLC Analysis of Chlorocresol on Chromolith® HighResolution RP-18 endcapped
column Chromolith® HighResolution RP-18 endcapped, 150 x 4.6 mm (1.52023.0001)
column temp. 30 °C
mobile phase [A] water; [B] acetonitrile; (55:45, A:B)
flow rate 1 mL/min
pressure 667 psi (46 Bar)
sample Dissolve 10 mg of chlorocresol standard with 100 mL of diluent
injection 10 μL
detector 240 nm cell: 10 μL
Featured Industry General Analytical
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany