Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Chlorpheniramine Enantiomers on Astec® CYCLOBOND® I 2000
HPLC Analysis of Chlorpheniramine Enantiomers on Astec® CYCLOBOND® I 2000
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. 23 °C
mobile phase [A] 1% triethylamine acetate, pH 4.1; [B] acetonitrile; (90:10, A:B)
flow rate 1 mL/min
sample 5 mg/mL in methanol
injection 2 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Antihistamines, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC