HPLC Analysis of Ciprofloxacin in Plasma on Chromolith(R) High Resolution RP-18e
HPLC Analysis of Ciprofloxacin in Plasma on Chromolith® High Resolution RP-18e
Conditions
column Chromolith High Resolution RP-18 endcapped, 100 x 4.6 mm (1.52022.0001)
column temp. ambient
mobile phase [A] methanol; [B] 20mM phosphat buffer pH=2.5: (15:85, A:B)
flow rate 3 mL/min
sample Ciprofloxacin 2,4 μg/mL solved in Albumin solution.
injection 3 μL
detector 277 nm cell: 11 μL
Description
Featured Industry Pharmaceutical (small molecule)
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany