HPLC Analysis of Cisplatin and Monohydrated Cisplatin on SeQuant ZIC-HILIC
HPLC Analysis of Cisplatin and Monohydrated Cisplatin on SeQuant ZIC-HILIC
Conditions
column Sequant ZIC-HILIC, 150 x 2.1 mm, 5 µm, 200 Å
mobile phase [A] 25 mM ammonium formate, pH 6.5; [B] 1,4-dioxane; (20:80, A:B)
flow rate 0.1 mL/min
column temp. ambient
detector UV, 305 nm
detector other analytical flow cell 1µL/10mm
injection 1 µL
Description
Analysis Note Antineoplastic agents separated with excellent resolution.
Featured Industry Pharmaceutical (small molecule)
Legal Information LiChrosolv is a trademark of Merck KGaA, Darmstadt, Germany
SeQuant is a trademark of Merck KGaA, Darmstadt, Germany
Application No. ASC140218C