Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Contents of Energy Drinks on Chromolith(R) Performance 100-3mm
HPLC Analysis of Contents of Energy Drinks on Chromolith® Performance 100-3mm
Conditions
sample preparation 1ml Sample on Extrelut N1, wait 10min., eluate with 6ml Ethylacetate, blow up Ethylacetate with nitrogen, dillute sample in 1ml water.
column Chromolith® Performance 100 x 3 mm (1.52001.0001)
column temp. 40 °C
mobile phase [A] methanol; [B] 20mM Phosphat buffer pH=5; (10:90, A:B)
gradient 0-3min 425 μl/min, 3,1-12min 850μl/min, 12,1-15min 425μl/min
sample Energy drink (Red Bull)
injection 1.8 μL
detector 227 nm cell: 1.4 μL
Description
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany