Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Contents of Energy Drinks with SPE on Chromolith(R) Performance 100 - 4.6 mm
HPLC Analysis of Contents of Energy Drinks with SPE on Chromolith® Performance 100 - 4.6 mm
Conditions
sample preparation 1ml Sample on Extrelut N1, wait 10min., eluate with 6ml Ethylacetate, blow up Ethylacetate with nitrogen, dillute sample in 1ml water.
column Chromolith® Performance 100 x 4.6 mm (1.5228.0001)
column temp. 40 °C
mobile phase [A] methanol; [B] 20mM Phosphat buffer pH=5; (10:90, A:B)
flow rate 0-3min 1mL/min, 3,1-12min 2,5mL/min, 12,1-15min 1mL/min
sample Energy drink (Red Bull)
injection 8 μL
detector 227 nm cell: 11 μL
Description
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany