HPLC Analysis of Contents of Energy Drinks without SPE on Chromolith(R) Performance 100 - 4.6 mm
HPLC Analysis of Contents of Energy Drinks without SPE on Chromolith® Performance 100 - 4.6 mm
Conditions
column Chromolith® Performance 100 x 4.6 mm (1.52028.0001)
column temp. 40 °C
mobile phase [A] methanol; [B] 20mM Phosphat buffer pH=5; (10:90, A:B)
flow rate 0-3min 1mL/min, 3,1-12min 2,5mL/min, 12,1-15min 1mL/min
sample Energy drink (Red Bull)
injection 8 μL
detector 227 nm cell: 11 μL
Description
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany