HPLC Analysis of Contents of Energy Drinks Without SPE on Chromolith(R) Performance 100-2mm
HPLC Analysis of Contents of Energy Drinks Without SPE on Chromolith® Performance 100-2mm
Conditions
sample preparation direct inject
column Chromolith® Performance 100 x 2 mm (1.52006.0001)
column temp. 40 °C
mobile phase [A] methanol; [B] 20mM Phosphat buffer pH=5; (10:90, A:B)
gradient 0-3min 190 μl/min, 3,1-12min 425μl/min, 12,1-15min 190μl/min
sample Energy drink (Red Bull)
injection 3 μL
detector 227 nm cell: 1.4 μL
Description
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany