Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Cysteine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)
HPLC Analysis of Cysteine Enantiomers on Astec® CHIROBIOTIC® T (Formic Acid Mobile Phase Additive)
Conditions
column Astec CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase [A] water; [B] methanol; [C] formic acid; (40:60:0.02, A:B:C)
flow rate 1 mL/min
sample (200 mug /mL in 50:50 water:methanol)
injection 10 µL
detector UV, 205 nm
Description
Categories Analytical Chromatography, Amino Acids
Featured Industry Life Science and Biopharma
Legal Information CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC, Astec is a registered trademark of Sigma-Aldrich Co. LLC