Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO PolyMA-SCX
HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO PolyMA-SCX
Conditions
column Discovery BIO PolyMA-SCX, 5 cm x 4.6 mm I.D., 5 µm particles (56901-U)
mobile phase [A] 20 mM Bis-Tris HCl, pH 7.0; [B] A + 0.5 M NaCl 24-69% B over 20 min
flow rate 0.5 mL/min
injection 10 mugeach variant
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC