HPLC Analysis of Deoxynivalenol (DON) on SeQuant ZIC-HILIC
HPLC Analysis of Deoxynivalenol (DON) on SeQuant ZIC-HILIC
Conditions
column Sequant ZIC-HILIC, PEEK, 25 cm x 4.6 mm, 5 µm, 200 Å
mobile phase   [A] acetonitrile; [B] ammonium acetate; (80:20, A,B)
column temp. ambient
detector UV, 220 nm
injection 10 µL
Description
Analysis Note HILIC mode separation provided rapid, sensitive determination under HPLC or LC/MS conditions.
Featured Industry Food & Beverage
Legal Information SeQuant is a trademark of Merck KGaA, Darmstadt, Germany
TraceCERT is a trademark of Sigma-Aldrich Co. LLC
Application No. ASC140218A